Assay Details
Target Gene Details
Entrez Gene ID: | 21987 |
Gene Name: | tumor protein D52-like 1 |
Gene Aliases: |
D53 |
Location: |
Chr.10:31332375-31446144 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpd52l1 | NM_009413.1 | NP_033439.1 | ||
| AF004428.1 | AAC98476.1 | |||
| AK131855.1 | BAE20832.1 | |||
| AK182630.1 | ||||
| BC141068.1 | ||||
| BC145321.1 |

Genomic Map