Assay Details
Target Gene Details
Entrez Gene ID: | 216343 |
Gene Name: | tryptophan hydroxylase 2 |
Gene Aliases: |
AU043594, Ntph |
Location: |
Chr.10:115078641-115185022 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tph2 | NM_173391.3 | 6 | 796 | NP_775567.2 |
| AY090565.1 | 6 | 796 | AAM08923.1 | |
| BC120514.1 | 6 | 787 | ||
| BC125430.1 | 6 | 787 |

Genomic Map