Assay Details
Target Gene Details
Entrez Gene ID: | 19707 |
Gene Name: | RalBP1 associated Eps domain containing protein |
Gene Aliases: |
BB161292 |
Location: |
Chr.10:18055561-18125155 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reps1 | NM_001111065.1 | 4 | 574 | NP_001104535.1 |
| NM_009048.2 | 4 | 574 | NP_033074.2 | |
| AF031939.1 | 3 | 395 | AAB94736.1 | |
| AK041967.1 | 2 | 1652 | BAC31117.1 | |
| AK150284.1 | 4 | 438 | BAE29439.1 | |
| AK151309.1 | 4 | 438 | BAE30291.1 | |
| BC087547.1 | 4 | 522 | AAH87547.1 |
More Information
Set Membership: |
|

Genomic Map