Assay Details
Target Gene Details
Entrez Gene ID: | 20222 |
Gene Name: | splicing factor 3a, subunit 2 |
Gene Aliases: |
66kDa, AW047242, PRP11, SFA66, Sap62 |
Location: |
Chr.10:80798664-80804924 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sf3a2 | NM_013651.4 | NP_038679.3 | ||
| BC052697.1 | AAH52697.1 | |||
| CX212776.1 |
More Information
Set Membership: |
|

Genomic Map