Assay Details
Target Gene Details
Entrez Gene ID: | 93759 |
Gene Name: | sirtuin 1 |
Gene Aliases: |
AA673258, SIR2L1, Sir2, Sir2a, Sir2alpha |
Location: |
Chr.10:63319005-63339061 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sirt1 | NM_001159589.1 | 6 | 1154 | NP_001153061.1 |
| NM_019812.2 | 7 | 1268 | NP_062786.1 | |
| AF214646.1 | 7 | 1253 | AAF24983.1 | |
| AK144203.1 | 6 | 1157 | BAE25766.1 | |
| AY377984.1 | 7 | 1253 | AAR23928.1 | |
| BC152314.1 | 7 | 748 |

Genomic Map