Assay Details
Target Gene Details
Entrez Gene ID: | 117150 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase, type II, gamma |
Gene Aliases: |
Pip5k2c |
Location: |
Chr.10:127197067-127211644 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip4k2c | NM_054097.3 | NP_473438.1 | ||
| AB054591.1 | BAB62096.1 | |||
| AK140079.1 | 3 | 475 | ||
| AK148299.1 | BAE28464.1 | |||
| AK182167.1 | ||||
| AK207886.1 | ||||
| BC021383.1 | AAH21383.1 |
More Information
Set Membership: |
|

Genomic Map