Assay Details
Target Gene Details
Entrez Gene ID: | 100038591 |
Gene Name: | RIKEN cDNA F420014N23 gene |
Gene Aliases: |
ENSMUSG00000074658 |
Location: |
Chr.10:127195249-127202643 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F420014N23Rik | NR_045715.1 | 4 | 1012 | |
| NR_045716.1 | 3 | 856 | ||
| AK165234.1 | 3 | 857 | BAE38092.1 |
Target Gene Details
Entrez Gene ID: | 117150 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase, type II, gamma |
Gene Aliases: |
Pip5k2c |
Location: |
Chr.10:127197067-127211644 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip4k2c | NM_054097.3 | 5 | 707 | NP_473438.1 |
| AB054591.1 | 5 | 650 | BAB62096.1 | |
| AK148299.1 | 5 | 667 | BAE28464.1 | |
| AK181991.1 | 2 | 118 | ||
| AK207886.1 | 3 | 315 | ||
| AK208052.1 | 2 | 120 | ||
| BC021383.1 | 5 | 640 | AAH21383.1 |
More Information
Set Membership: |
|

Genomic Map