Assay Details
Target Gene Details
Entrez Gene ID: | 100038591 |
Gene Name: | RIKEN cDNA F420014N23 gene |
Gene Aliases: |
ENSMUSG00000074658 |
Location: |
Chr.10:127195249-127202643 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F420014N23Rik | NR_045715.1 | |||
| NR_045716.1 | ||||
| AK077682.1 | ||||
| AK165234.1 | BAE38092.1 |
Target Gene Details
Entrez Gene ID: | 117150 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase, type II, gamma |
Gene Aliases: |
Pip5k2c |
Location: |
Chr.10:127197067-127211644 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip4k2c | NM_054097.3 | NP_473438.1 | ||
| AB054591.1 | BAB62096.1 | |||
| AK148299.1 | BAE28464.1 | |||
| AK208052.1 | ||||
| BC005539.1 | AAH05539.1 | |||
| BC021383.1 | AAH21383.1 |
More Information
Set Membership: |
|

Genomic Map