Assay Details
Target Gene Details
Entrez Gene ID: | 170728 |
Gene Name: | reticulon 4 interacting protein 1 |
Gene Aliases: |
D10Ertd690e, NIMP |
Location: |
Chr.10:43901807-43947862 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtn4ip1 | NM_130892.4 | 7 | 1354 | NP_570962.2 |
| AF336862.1 | 7 | 1084 | AAK64604.1 | |
| AK050324.1 | 7 | 1352 | BAC34189.1 | |
| AK085858.1 | 7 | 1334 | BAC39556.1 | |
| AK088029.1 | 7 | 1355 | BAC40106.1 | |
| BC024116.1 | 7 | 1324 | AAH24116.1 |
More Information
Set Membership: |
|

Genomic Map