Assay Details
Target Gene Details
Entrez Gene ID: | 20847 |
Gene Name: | signal transducer and activator of transcription 2 |
Gene Aliases: |
1600010G07Rik, AW496480 |
Location: |
Chr.10:128270559-128292852 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat2 | NM_019963.1 | 21 | 2138 | NP_064347.1 |
| AF088862.1 | 21 | 2011 | AAD38329.1 | |
| AF187231.1 | 21 | 2135 | AAF01441.1 | |
| AF206162.1 | 21 | 2138 | AAF17096.1 | |
| AK030903.1 | 14 | 1230 | BAC27177.1 | |
| AK149924.1 | 21 | 2151 | BAE29170.1 | |
| AK150070.1 | 21 | 2150 | BAE29282.1 | |
| AK153405.1 | 19 | 1785 | BAE31965.1 | |
| BC064827.1 | 10 | 1135 | AAH64827.1 |
More Information
Set Membership: |
|

Genomic Map