Assay Details
Target Gene Details
Entrez Gene ID: | 20847 |
Gene Name: | signal transducer and activator of transcription 2 |
Gene Aliases: |
1600010G07Rik, AW496480 |
Location: |
Chr.10:128270559-128292852 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat2 | NM_019963.1 | NP_064347.1 | ||
| AF088862.1 | AAD38329.1 | |||
| AF187231.1 | AAF01441.1 | |||
| AF206162.1 | AAF17096.1 | |||
| AK149838.1 | ||||
| AK149924.1 | BAE29170.1 | |||
| AK150070.1 | BAE29282.1 | |||
| AK153405.1 | BAE31965.1 |
More Information
Set Membership: |
|

Genomic Map