Assay Details
Target Gene Details
Entrez Gene ID: | 18717 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase, type 1 gamma |
Gene Aliases: |
AI115456, AI835305, Pip5kIgamma |
Location: |
Chr.10:81292962-81319974 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip5k1c | NM_001146687.2 | 12 | 1476 | NP_001140159.1 |
| NM_001293646.1 | 12 | 1476 | NP_001280575.1 | |
| NM_001293647.1 | 12 | 1489 | NP_001280576.1 | |
| NM_008844.3 | 12 | 1476 | NP_032870.2 | |
| AB006916.1 | 12 | 1459 | BAA25664.1 | |
| AK014403.1 | 12 | 1442 | ||
| AK041402.1 | 12 | 1462 | ||
| AK075823.1 | ||||
| AK122319.2 | 12 | 1434 | BAC65601.2 | |
| AK154816.1 | 12 | 1468 | BAE32849.1 | |
| AK155371.1 | 12 | 1452 | ||
| AK170286.1 | 12 | 1468 | ||
| AK171576.1 | 12 | 1450 | BAE42536.1 | |
| AK194542.1 | ||||
| BC019138.1 | 12 | 1425 | AAH19138.1 | |
| BC094665.1 | 12 | 1448 | AAH94665.1 |
More Information
Set Membership: |
|

Genomic Map