Assay Details
Target Gene Details
Entrez Gene ID: | 19156 |
Gene Name: | prosaposin |
Gene Aliases: |
AI037048, SGP-1 |
Location: |
Chr.10:60277628-60302600 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 19156 |
Gene Name: | prosaposin |
Gene Aliases: |
AI037048, SGP-1 |
Location: |
Chr.10:60277628-60302600 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Set Membership: |
|