Assay Details
Target Gene Details
Entrez Gene ID: | 76563 |
Gene Name: | glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1 |
Gene Aliases: |
2700038P16Rik, C80053, GatA |
Location: |
Chr.10:43874188-43901739 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qrsl1 | NM_001081054.2 | NP_001074523.1 | ||
| AK012351.1 | BAB28181.1 | |||
| AK038801.1 | ||||
| AK041347.1 | ||||
| AK170225.1 | BAE41646.1 | |||
| BC070459.1 | AAH70459.1 |
More Information
Set Membership: |
|

Genomic Map