Assay Details
Target Gene Details
Entrez Gene ID: | 66932 |
Gene Name: | REX1, RNA exonuclease 1 homolog (S. cerevisiae) |
Gene Aliases: |
1700021P10Rik, 2610511M11Rik, AW050347, EloA-BP1, Tceb3bp1 |
Location: |
Chr.10:80540926-80563381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rexo1 | NM_025852.3 | NP_080128.2 | ||
| AK006228.1 | BAB24471.1 | |||
| AK007009.1 | ||||
| AK079120.1 | ||||
| AK173108.1 | BAD32386.1 | |||
| BC049901.1 | AAH49901.1 | |||
| BC052424.1 | AAH52424.1 |
More Information
Set Membership: |
|

Genomic Map