Assay Details
Target Gene Details
Entrez Gene ID: | 70432 |
Gene Name: | RUN and FYVE domain-containing 2 |
Gene Aliases: |
2610111M19Rik, AI844453, Denn, LZ-FYVE, ZFYVE13 |
Location: |
Chr.10:62980223-63018742 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rufy2 | NM_027425.3 | 10 | 979 | NP_081701.2 |
| AK011847.1 | 10 | 949 | BAB27878.1 | |
| AK083205.1 | 10 | 964 | BAC38807.1 | |
| AK173195.1 | 10 | 940 | BAD32473.1 | |
| BC017645.1 | 10 | 941 | ||
| BC137736.1 | 10 | 928 | ||
| BC137737.1 | 10 | 928 |

Genomic Map