Assay Details
Target Gene Details
Entrez Gene ID: | 70432 |
Gene Name: | RUN and FYVE domain-containing 2 |
Gene Aliases: |
2610111M19Rik, AI844453, Denn, LZ-FYVE, ZFYVE13 |
Location: |
Chr.10:62980223-63018742 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rufy2 | NM_027425.3 | NP_081701.2 | ||
| AK011847.1 | BAB27878.1 | |||
| AK045485.1 | 3 | 431 | ||
| AK083205.1 | BAC38807.1 | |||
| AK173195.1 | BAD32473.1 | |||
| BB643897.1 | 3 | 431 | ||
| BC017645.1 | ||||
| BC137736.1 | ||||
| BC137737.1 |

Genomic Map