Assay Details
Target Gene Details
Entrez Gene ID: | 216049 |
Gene Name: | zinc finger protein 365 |
Gene Aliases: |
AI839779, AV340892, DBZ, Su48, Znf365, talanin |
Location: |
Chr.10:67886105-67912662 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp365 | NM_178679.2 | 3 | 1187 | NP_848794.1 |
| AF543761.1 | 2 | 908 | AAQ11828.1 | |
| AJ516033.1 | 3 | 1049 | CAD56774.1 | |
| AK031846.1 | 3 | 983 | BAC27578.1 | |
| AK033892.1 | 3 | 949 | BAC28505.1 | |
| AK044322.1 | 3 | 946 | BAC31866.1 | |
| AK044655.1 | 2 | 270 | BAC32019.1 | |
| AK077316.1 | 3 | 960 | BAC36745.1 | |
| AK122387.1 | 3 | 1174 | BAC65669.1 | |
| BC080272.1 | 3 | 1147 | AAH80272.1 | |
| BC157983.1 | 3 | 988 | ||
| BC157991.1 | 3 | 988 |

Genomic Map