Assay Details
Target Gene Details
Entrez Gene ID: | 72503 |
Gene Name: | RIKEN cDNA 2610507B11 gene |
Gene Aliases: |
AU022687, D11Bhm178e, D11Bhm179e, E1, Kiaa0100, mKIAA0100 |
Location: |
Chr.11:78261703-78290625 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610507B11Rik | NM_001002004.2 | 4 | 420 | NP_001002004.2 |
| AK149719.1 | 4 | 391 | BAE29046.1 | |
| AK150829.1 | 4 | 421 | ||
| BB866513.1 | 4 | 420 | ||
| BC019798.1 | 4 | 373 | ||
| BC075633.1 | 4 | 380 | AAH75633.1 | |
| BC080706.1 | 4 | 397 | AAH80706.1 |

Genomic Map