Assay Details
Target Gene Details
Entrez Gene ID: | 216049 |
Gene Name: | zinc finger protein 365 |
Gene Aliases: |
AI839779, AV340892, DBZ, Su48, Znf365, talanin |
Location: |
Chr.10:67886105-67912662 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp365 | NM_178679.2 | NP_848794.1 | ||
| AF543761.1 | AAQ11828.1 | |||
| AJ516033.1 | CAD56774.1 | |||
| AK031846.1 | BAC27578.1 | |||
| AK033892.1 | BAC28505.1 | |||
| AK044322.1 | BAC31866.1 | |||
| AK044655.1 | BAC32019.1 | |||
| AK122387.1 | BAC65669.1 | |||
| BC080272.1 | AAH80272.1 | |||
| BC157983.1 | ||||
| BC157991.1 |

Genomic Map