Assay Details
Target Gene Details
Entrez Gene ID: | 103406 |
Gene Name: | zinc finger RNA binding protein 2 |
Gene Aliases: |
2010013I23Rik, 9130206N08Rik, AW045600, mKIAA1086 |
Location: |
Chr.10:81233163-81252123 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfr2 | NM_001034895.3 | 16 | 2316 | NP_001030067.2 |
| AK033649.1 | 9 | 1065 | ||
| AK122438.1 | 7 | 4434 | BAC65720.3 | |
| BQ042879.1 | 4 | 337 |
More Information
Set Membership: |
|

Genomic Map