Assay Details
Target Gene Details
Entrez Gene ID: | 237754 |
Gene Name: | butyrophilin-like 9 |
Gene Aliases: |
B430208I01, Btn3, D330012D11Rik |
Location: |
Chr.11:49168325-49187089 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btnl9 | NM_172793.2 | 2 | 174 | NP_766381.1 |
| AK080918.1 | 2 | 157 | ||
| AK084528.1 | 2 | 175 | BAC39210.1 | |
| AY177687.1 | 2 | 141 | AAO38197.1 | |
| BC025540.1 | 2 | 152 | AAH25540.1 | |
| BC150808.1 | 2 | 131 | ||
| BC151155.1 | 2 | 131 |
More Information
Set Membership: |
|

Genomic Map