Assay Details
Target Gene Details
Entrez Gene ID: | 12295 |
Gene Name: | calcium channel, voltage-dependent, beta 1 subunit |
Gene Aliases: |
CAB1, Cchb1, Cchlb1 |
Location: |
Chr.11:98001508-98022975 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacnb1 | NM_001159319.2 | NP_001152791.1 | ||
| NM_001282977.1 | NP_001269906.1 | |||
| NM_001282978.1 | NP_001269907.1 | |||
| NM_031173.4 | NP_112450.1 | |||
| NM_145121.3 | NP_660099.2 | |||
| AB685743.1 | ||||
| AF068898.1 | AAC19386.1 | |||
| AF322905.1 | AAG42829.1 | |||
| AK155193.1 | BAE33107.1 | |||
| AY094172.1 | AAM11473.1 | |||
| AY094173.1 | AAM11474.1 | |||
| BC077713.1 | AAH77713.1 | |||
| BY080419.1 | ||||
| CA316737.1 |
More Information
Set Membership: |
|

Genomic Map