Assay Details
Target Gene Details
Entrez Gene ID: | 103406 |
Gene Name: | zinc finger RNA binding protein 2 |
Gene Aliases: |
2010013I23Rik, 9130206N08Rik, AW045600, mKIAA1086 |
Location: |
Chr.10:81233163-81252123 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfr2 | NM_001034895.3 | 2 | 236 | NP_001030067.2 |
| AK165530.1 | 2 | 224 | BAE38241.1 | |
| BB641594.1 | 2 | 236 | ||
| CB249573.1 | 2 | 212 |
More Information
Set Membership: |
|

Genomic Map