Assay Details
Target Gene Details
Entrez Gene ID: | 11421 |
Gene Name: | angiotensin I converting enzyme (peptidyl-dipeptidase A) 1 |
Gene Aliases: |
AW208573, CD143 |
Location: |
Chr.11:105967945-105989964 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ace | NM_001281819.1 | 20 | 3032 | NP_001268748.1 |
| NM_009598.2 | 9 | 1255 | NP_033728.1 | |
| NM_207624.5 | 20 | 3032 | NP_997507.1 | |
| AK154632.1 | 20 | 3031 | BAE32728.1 | |
| AK161020.1 | 20 | 3028 | BAE36151.1 | |
| AK164780.1 | 7 | 900 | ||
| BC034367.1 | 16 | 2294 | AAH34367.1 | |
| BC040404.1 | 20 | 3021 | AAH40404.1 | |
| BC083109.1 | 20 | 2810 | AAH83109.1 | |
| BC110362.1 | 20 | 3018 | AAI10363.1 | |
| J04946.1 | 14 | 1999 | AAA37147.1 | |
| M55333.1 | 9 | 1255 | AAA37149.1 |
Target Gene Details
Entrez Gene ID: | 100379610 |
Gene Name: | predicted gene 11651 |
Gene Aliases: |
OTTMUSG00000003114 |
Location: |
Chr.11:105965607-105985694 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm11651 | NR_104047.1 | |||
| AA145017.1 | ||||
| AK016887.1 | ||||
| CF106408.1 |
More Information
Set Membership: |
|

Genomic Map