Assay Details
Target Gene Details
Entrez Gene ID: | 11421 |
Gene Name: | angiotensin I converting enzyme (peptidyl-dipeptidase A) 1 |
Gene Aliases: |
AW208573, CD143 |
Location: |
Chr.11:105967945-105989964 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ace | NM_001281819.1 | 24 | 3643 | NP_001268748.1 |
| NM_009598.2 | 13 | 1866 | NP_033728.1 | |
| NM_207624.5 | 24 | 3643 | NP_997507.1 | |
| AK087056.1 | BAC39791.1 | |||
| AK154632.1 | 24 | 3642 | BAE32728.1 | |
| AK161020.1 | 24 | 3639 | BAE36151.1 | |
| AK164780.1 | 11 | 1511 | ||
| BC034367.1 | 20 | 2905 | AAH34367.1 | |
| BC040404.1 | 24 | 3632 | AAH40404.1 | |
| BC052531.1 | 5 | 495 | AAH52531.1 | |
| BC083109.1 | 24 | 3421 | AAH83109.1 | |
| BC110362.1 | 24 | 3629 | AAI10363.1 | |
| J04946.1 | 18 | 2610 | AAA37147.1 | |
| M55333.1 | 13 | 1866 | AAA37149.1 |
More Information
Set Membership: |
|

Genomic Map