Assay Details
Target Gene Details
Entrez Gene ID: | 216869 |
Gene Name: | arrestin, beta 2 |
Gene Aliases: |
AI326910, AW122872 |
Location: |
Chr.11:70432580-70440828 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arrb2 | NM_001271358.1 | NP_001258287.1 | ||
| NM_001271359.1 | NP_001258288.1 | |||
| NM_001271360.1 | NP_001258289.1 | |||
| NM_145429.5 | NP_663404.1 | |||
| AK089542.1 | ||||
| AK154874.1 | BAE32894.1 | |||
| AK159317.1 | BAE34984.1 | |||
| AK162889.1 | BAE37102.1 | |||
| AK170647.1 | BAE41934.1 | |||
| AK170889.1 | BAE42096.1 | |||
| BC016642.1 | AAH16642.1 |
More Information
Set Membership: |
|

Genomic Map