Assay Details
Target Gene Details
Entrez Gene ID: | 12299 |
Gene Name: | calcium channel, voltage-dependent, gamma subunit 1 |
Gene Aliases: |
- |
Location: |
Chr.11:107703218-107716476 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacng1 | NM_007582.2 | NP_031608.1 | ||
| AJ006306.1 | CAA06966.1 | |||
| AK132580.1 | BAE21242.1 | |||
| BC099482.1 | AAH99482.1 |
More Information
Set Membership: |
|

Genomic Map