Assay Details
Target Gene Details
Entrez Gene ID: | 327799 |
Gene Name: | ubiquitin specific peptidase 44 |
Gene Aliases: |
E430004F17Rik |
Location: |
Chr.10:93819968-93860738 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp44 | NM_001206851.1 | NP_001193780.1 | ||
| NM_183199.3 | NP_899022.2 | |||
| AK088109.1 | BAC40148.1 |
More Information
Set Membership: |
|

Genomic Map