Assay Details
Target Gene Details
Entrez Gene ID: | 52696 |
Gene Name: | ZW10 interactor |
Gene Aliases: |
2010007E07Rik, 2600001N01Rik, D10Ertd749e, Zwint-1 |
Location: |
Chr.10:72654846-72674964 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zwint | NM_001293683.1 | NP_001280612.1 | ||
| NM_001293684.1 | NP_001280613.1 | |||
| NM_025635.4 | NP_079911.1 | |||
| AK004305.1 | BAB23256.1 | |||
| AK008144.1 | BAB25492.1 | |||
| AK013556.1 | BAB28904.1 | |||
| AK075977.1 | BAC36090.1 | |||
| AK082215.1 | ||||
| AK134380.1 | BAE22123.1 | |||
| AK170637.1 | ||||
| BC013559.1 | AAH13559.1 | |||
| BC027100.1 | AAH27100.1 | |||
| BC034870.1 | AAH34870.1 |

Genomic Map