Assay Details
Target Gene Details
Entrez Gene ID: | 11749 |
Gene Name: | annexin A6 |
Gene Aliases: |
AW107198, Anx6, AnxVI, Cabm, Camb |
Location: |
Chr.11:54978962-55033471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anxa6 | NM_001110211.1 | NP_001103681.1 | ||
| NM_013472.4 | NP_038500.2 | |||
| AK010342.1 | BAC25291.1 | |||
| AK013026.1 | ||||
| AK030728.1 | BAC27101.1 | |||
| AK077843.1 | BAE43362.1 | |||
| AK087941.1 | ||||
| AK146509.1 | BAE27222.1 | |||
| AK146592.1 | BAE27287.1 | |||
| AK147066.1 | BAE27650.1 | |||
| AK150035.1 | BAE29257.1 | |||
| AK160754.1 | BAE35990.1 | |||
| AK169774.1 | ||||
| AK217048.1 | ||||
| BC005595.1 | AAH05595.1 | |||
| X13460.1 | CAA31808.1 |
More Information
Set Membership: |
|

Genomic Map