Assay Details
Target Gene Details
Entrez Gene ID: | 74008 |
Gene Name: | arylsulfatase G |
Gene Aliases: |
6330406P08Rik, AI846872 |
Location: |
Chr.11:109450845-109573330 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsg | NM_028710.3 | 8 | 1494 | NP_082986.3 |
| AK018132.2 | 5 | 911 | BAB31086.1 | |
| AK158726.1 | 8 | 1495 | BAE34629.1 | |
| AK173082.1 | 5 | 593 | BAD32360.1 | |
| AK192752.1 | 4 | 242 | ||
| BC022158.1 | 4 | 324 | AAH22158.1 | |
| BC084731.1 | 5 | 1184 | AAH84731.1 |
More Information
Set Membership: |
|

Genomic Map