Assay Details
Target Gene Details
Entrez Gene ID: | 74008 |
Gene Name: | arylsulfatase G |
Gene Aliases: |
6330406P08Rik, AI846872 |
Location: |
Chr.11:109450845-109573330 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsg | NM_028710.3 | 9 | 1663 | NP_082986.3 |
| AK018132.2 | 6 | 1080 | BAB31086.1 | |
| AK158726.1 | 9 | 1664 | BAE34629.1 | |
| AK173082.1 | 6 | 762 | BAD32360.1 | |
| AK192114.1 | 1 | 69 | ||
| AK192752.1 | ||||
| BC022158.1 | 5 | 493 | AAH22158.1 | |
| BC084731.1 | 6 | 1353 | AAH84731.1 |
More Information
Set Membership: |
|

Genomic Map