Assay Details
Target Gene Details
Entrez Gene ID: | 74008 |
Gene Name: | arylsulfatase G |
Gene Aliases: |
6330406P08Rik, AI846872 |
Location: |
Chr.11:109450845-109573330 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsg | NM_001166177.1 | NP_001159649.1 | ||
| NM_028710.3 | NP_082986.3 | |||
| AK018132.2 | BAB31086.1 | |||
| AK158726.1 | BAE34629.1 | |||
| AK173082.1 | BAD32360.1 | |||
| AK186315.1 | ||||
| AK192114.1 | ||||
| BC022158.1 | AAH22158.1 | |||
| BC039629.1 | AAH39629.1 | |||
| BC084731.1 | AAH84731.1 | |||
| BY005671.1 | ||||
| BY095625.1 |
More Information
Set Membership: |
|

Genomic Map