Assay Details
Target Gene Details
Entrez Gene ID: | 11975 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit A1 |
Gene Aliases: |
AA959968, ATP6a1, Atp6n1, Atp6n1a, Atpv0a1, Vpp-1, Vpp1 |
Location: |
Chr.11:101009447-101063719 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0a1 | NM_001243049.1 | NP_001229978.1 | ||
| NM_001243050.1 | 2 | 161 | NP_001229979.1 | |
| NM_001243051.1 | 2 | 161 | NP_001229980.1 | |
| NM_016920.3 | 2 | 161 | NP_058616.1 | |
| AB022321.1 | BAA93005.1 | |||
| AF218249.1 | 2 | 131 | AAF59918.1 | |
| AF218250.1 | AAF59919.1 | |||
| AF218251.1 | AAF59920.1 | |||
| AK029316.1 | 2 | 133 | ||
| AK036530.1 | 2 | 130 | ||
| AK040343.1 | 2 | 69 | BAE20575.1 | |
| AK158786.1 | BAE34665.1 | |||
| AK159114.1 | 2 | 97 | BAE34830.1 | |
| AK172078.1 | 2 | 92 | ||
| AK192836.1 | ||||
| AK195947.1 | ||||
| AK198173.1 | ||||
| AK204666.1 | ||||
| AK214847.1 | 1 | 57 | ||
| AK215374.1 | ||||
| AW318708.1 | 2 | 91 | ||
| BC001995.1 | 2 | 127 | AAH01995.1 | |
| BC066839.1 | 2 | 126 | AAH66839.1 | |
| BC071182.1 | 2 | 126 | AAH71182.1 | |
| BY207073.1 | 2 | 159 | ||
| CJ143241.1 | 2 | 162 | ||
| U13836.1 | AAC83083.1 |
More Information
Set Membership: |
|

Genomic Map