Assay Details
Target Gene Details
Entrez Gene ID: | 11975 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit A1 |
Gene Aliases: |
AA959968, ATP6a1, Atp6n1, Atp6n1a, Atpv0a1, Vpp-1, Vpp1 |
Location: |
Chr.11:101009447-101063719 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0a1 | NM_001243049.1 | NP_001229978.1 | ||
| NM_001243050.1 | NP_001229979.1 | |||
| NM_001243051.1 | NP_001229980.1 | |||
| NM_016920.3 | NP_058616.1 | |||
| AB022321.1 | BAA93005.1 | |||
| AF218249.1 | AAF59918.1 | |||
| AF218250.1 | AAF59919.1 | |||
| AF218251.1 | AAF59920.1 | |||
| AK029316.1 | ||||
| AK158786.1 | BAE34665.1 | |||
| AK159114.1 | BAE34830.1 | |||
| AK172078.1 | ||||
| AK181497.1 | ||||
| AK196945.1 | ||||
| AK200735.1 | ||||
| BC066839.1 | AAH66839.1 | |||
| BC071182.1 | AAH71182.1 | |||
| U13836.1 | AAC83083.1 |
More Information
Set Membership: |
|

Genomic Map