Assay Details
Target Gene Details
Entrez Gene ID: | 12261 |
Gene Name: | complement component 1, q subcomponent binding protein |
Gene Aliases: |
AA407365, AA986492, D11Wsu182e, HABP1, P32, gC1qBP |
Location: |
Chr.11:70977846-70983026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1qbp | NM_007573.2 | NP_031599.2 | ||
| AF300619.1 | AAL77246.1 | |||
| AJ001101.1 | CAA04530.1 | |||
| AK010601.1 | ||||
| AK010746.1 | ||||
| AK011868.1 | ||||
| AK169764.1 | ||||
| BC038075.1 | AAH38075.1 |

Genomic Map