Assay Details
Target Gene Details
Entrez Gene ID: | 12323 |
Gene Name: | calcium/calmodulin-dependent protein kinase II, beta |
Gene Aliases: |
- |
Location: |
Chr.11:5969642-6065748 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2b | NM_001174053.1 | NP_001167524.1 | ||
| NM_001174054.1 | NP_001167525.1 | |||
| NM_007595.5 | NP_031621.3 | |||
| AK046454.1 | BAC32736.1 | |||
| AK158794.1 | BAE34670.1 | |||
| AK163698.1 | BAE37464.1 | |||
| BC080273.1 | AAH80273.1 | |||
| X63615.1 | CAA45160.1 |
More Information
Set Membership: |
|

Genomic Map