Assay Details
Target Gene Details
Entrez Gene ID: | 382406 |
Gene Name: | POC1 centriolar protein homolog B (Chlamydomonas) |
Gene Aliases: |
4933430F16Rik, Wdr51b |
Location: |
Chr.10:99107171-99198074 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Poc1b | NM_027740.6 | NP_082016.1 | ||
| AK045302.1 | BAC32302.1 | |||
| AK050233.1 | BAC34138.1 | |||
| AK090075.1 | BAC41080.1 | |||
| BC145801.1 |
More Information
Set Membership: |
|

Genomic Map