Assay Details
Target Gene Details
Entrez Gene ID: | 66179 |
Gene Name: | 2-oxoglutarate and iron-dependent oxygenase domain containing 3 |
Gene Aliases: |
1110031I02Rik |
Location: |
Chr.11:121177593-121204648 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogfod3 | NM_025402.2 | 2 | 315 | NP_079678.1 |
| AK004005.1 | 2 | 316 | BAB23119.1 | |
| AK007883.1 | 2 | 377 | ||
| AK145938.1 | 2 | 380 | BAE26768.1 | |
| AK168300.1 | 2 | 351 | BAE40241.1 | |
| BC024349.1 | 2 | 282 | AAH24349.1 |
More Information
Set Membership: |
|

Genomic Map