Assay Details
Target Gene Details
Entrez Gene ID: | 72046 |
Gene Name: | upregulator of cell proliferation |
Gene Aliases: |
2010005J08Rik, 9130001I21Rik, AW060359, Urg4, mKIAA1507 |
Location: |
Chr.11:5713417-5762436 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Urgcp | NM_001077661.1 | NP_001071129.1 | ||
| NM_178623.3 | NP_848738.2 | |||
| AK033577.1 | BAC28370.1 | |||
| AK035580.1 | BAC29111.1 | |||
| AK041953.1 | BAC31109.1 | |||
| AK129378.1 | BAC98188.1 | |||
| AK166495.1 | BAE38807.1 | |||
| AK190812.1 | ||||
| AK191392.1 | ||||
| AK211020.1 | ||||
| BC066994.1 | AAH66994.1 | |||
| BY058620.1 |
More Information
Set Membership: |
|

Genomic Map