Assay Details
Target Gene Details
Entrez Gene ID: | 668433 |
Gene Name: | RIKEN cDNA 4930544D05 gene |
Gene Aliases: |
- |
Location: |
Chr.11:70615834-70616892 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930544D05Rik | NM_001145537.1 | NP_001139009.1 | ||
| AK019734.1 |
Target Gene Details
Entrez Gene ID: | 11448 |
Gene Name: | cholinergic receptor, nicotinic, epsilon polypeptide |
Gene Aliases: |
AChrepsilon, Acre, nAChRE |
Location: |
Chr.11:70614883-70619835 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrne | NM_009603.1 | NP_033733.1 | ||
| X55718.1 | CAA39251.1 |
More Information
Set Membership: |
|

Genomic Map