Assay Details
Target Gene Details
Entrez Gene ID: | 11890 |
Gene Name: | asialoglycoprotein receptor 2 |
Gene Aliases: |
ASGPR2, Asgr, Asgr-2 |
Location: |
Chr.11:70092307-70106187 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asgr2 | NM_007493.3 | NP_031519.1 | ||
| BC011197.1 | AAH11197.1 | |||
| X53042.1 | CAA37211.1 |
More Information
Set Membership: |
|

Genomic Map