Assay Details
Target Gene Details
Entrez Gene ID: | 216976 |
Gene Name: | cDNA sequence BC030499 |
Gene Aliases: |
BC037104, Sgk494 |
Location: |
Chr.11:78290732-78296805 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC030499 | NM_001287206.1 | NP_001274135.1 | ||
| AK138440.1 | ||||
| BY266382.1 |
More Information
Set Membership: |
|

Genomic Map