Assay Details
Target Gene Details
Entrez Gene ID: | 237711 |
Gene Name: | echinoderm microtubule associated protein like 6 |
Gene Aliases: |
2900083P10Rik, C230094A16Rik, EMAP-6 |
Location: |
Chr.11:29742586-30026034 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eml6 | NM_146016.2 | 24 | 4018 | NP_666128.2 |
| AK049034.1 | 24 | 3930 |

Genomic Map