Assay Details
Target Gene Details
Entrez Gene ID: | 66180 |
Gene Name: | prolyl 3-hydroxylase family member 4 (non-enzymatic) |
Gene Aliases: |
1110036O03Rik, AI413214, Leprel4, Sc65 |
Location: |
Chr.11:100408748-100414819 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3h4 | NM_176830.2 | 7 | 1399 | NP_789800.1 |
| AK029271.1 | 7 | 1279 | BAC26365.1 | |
| BC031856.1 | 7 | 1399 | AAH31856.1 |
More Information
Set Membership: |
|

Genomic Map