Assay Details
Target Gene Details
Entrez Gene ID: | 214604 |
Gene Name: | RIKEN cDNA 4932411E22 gene |
Gene Aliases: |
- |
Location: |
Chr.11:89390223-89418948 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4932411E22Rik | NM_172534.2 | 3 | 331 | NP_766122.1 |
| AK029964.1 | 3 | 332 | BAC26702.1 | |
| BC140961.1 | 3 | 317 |

Genomic Map