Assay Details
Target Gene Details
Entrez Gene ID: | 328019 |
Gene Name: | spermatogenesis associated 32 |
Gene Aliases: |
4933400C05Rik, VAD1.2 |
Location: |
Chr.11:103208127-103218432 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata32 | NM_177801.3 | NP_808469.1 | ||
| AK077068.1 | BAC36591.1 | |||
| BC117072.1 | ||||
| BC120775.1 | ||||
| DQ118647.1 | AAZ38360.1 |

Genomic Map