Assay Details
Target Gene Details
Entrez Gene ID: | 217265 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 5 |
Gene Aliases: |
ABC13, B930033A02Rik, mKIAA1888 |
Location: |
Chr.11:110269369-110337724 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca5 | NM_147219.2 | 19 | 2780 | NP_671752.2 |
| AB097675.1 | 18 | 2530 | BAC66658.1 | |
| AF491842.1 | 19 | 2739 | AAM90895.1 | |
| AK034961.1 | 1 | 136 | BAC28896.2 | |
| AK046203.1 | 20 | 2777 | ||
| AK047188.1 | 19 | 2730 | BAC32984.1 | |
| AK138604.1 | 19 | 2597 | BAE23713.1 | |
| BC064823.1 | 19 | 2742 | AAH64823.1 |
More Information
Set Membership: |
|

Genomic Map