Assay Details
Target Gene Details
Entrez Gene ID: | 216831 |
Gene Name: | Rho GTPase activating protein 44 |
Gene Aliases: |
6330543G20, AI840762, AU040829, AW493732, Rich2 |
Location: |
Chr.11:65002039-65163120 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap44 | NM_001099288.1 | 17 | 1919 | NP_001092758.1 |
| NM_175003.3 | 17 | 1919 | NP_778168.2 | |
| AK079081.1 | 13 | 1238 | ||
| BC056366.1 | 17 | 1919 | AAH56366.1 | |
| BC059911.1 | 17 | 1919 | AAH59911.1 |

Genomic Map